siRNA Details
| Gene Symbol: B230217C12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018341.1 | - | 2 | 1369 | ||
| AK031146.1 | - | 1368 | ||||
| AK035650.1 | - | 1351 | ||||
| AK042827.1 | - | 1371 | ||||
| AK080808.1 | - | 1368 | ||||
| AK081899.1 | - | 1389 | ||||

B230217C12Rik
Genomic Map