siRNA Details
| Gene Symbol: 4930543I11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016030.1 | - | 1 | 948 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV269527.1 | - | 1 | 258 | Chr. 10 - 67786422 - 67786695 [+] on Build GRCm38 | |

4930543I11Rik
Genomic Map