siRNA Details
| Gene Symbol: Gm15417 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040403.1 | 2 | 1155 | |||
| NR_040404.1 | 2 | 616 | ||||
| GenBank mRNA | AK136654.1 | - | 2 | 617 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CX210895.1 | - | 1 | 289 | Chr. 3 - 89398182 - 89398777 [+] on Build GRCm38 | |

Gm15417
Genomic Map