siRNA Details
| Gene Symbol: Msantd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013318.1 | - | 1 | 515 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Msantd2| Gene Symbol: Msantd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013318.1 | - | 1 | 515 | ||