siRNA Details
Gene Symbol: 5430427M07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045858.1 | 4 | 967 | |||
GenBank mRNA | AK017349.1 | - | 4 | 967 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 5430427M07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045858.1 | 4 | 967 | |||
GenBank mRNA | AK017349.1 | - | 4 | 967 |