siRNA Details
| Gene Symbol: 5430427G11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017348.1 | - | 1 | 625 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY717162.1 | - | 1 | 624 | Chr. 10 - 43220402 - 43221382 [-] on Build GRCm38 | |

5430427G11Rik
Genomic Map