siRNA Details
| Gene Symbol: 1700024I08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006304.1 | - | 3 | 363 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706381.1 | - | 3 | 360 | Chr. 13 - 63675768 - 63734298 [-] on Build GRCm38 | |

1700024I08Rik
Genomic Map