siRNA Details
| Gene Symbol: 1700020K04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006169.1 | - | 1 | 380 | ||
| AV204637.1 | - | 1 | 200 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706270.1 | - | 1 | 377 | Chr. 10 - 63800871 - 63801353 [+] on Build GRCm38 | |

1700020K04Rik
Genomic Map