siRNA Details
| Gene Symbol: C530043K16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK083074.1 | - | 1 | 1515 | ||
| AK084923.1 | - | 1 | 1506 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C530043K16Rik| Gene Symbol: C530043K16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK083074.1 | - | 1 | 1515 | ||
| AK084923.1 | - | 1 | 1506 | |||