siRNA Details
| Gene Symbol: Gm26664 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK037841.1 | - | 1 | 133 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB631112.1 | - | 1 | 133 | Chr. 8 - 84665128 - 84669336 [+] on Build GRCm38 | |

Gm26664
Genomic Map