siRNA Details
Gene Symbol: 2410088K16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040493.1 | 2 | 265 | |||
GenBank mRNA | AK019125.1 | - | 2 | 267 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 2410088K16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040493.1 | 2 | 265 | |||
GenBank mRNA | AK019125.1 | - | 2 | 267 |