siRNA Details
| Gene Symbol: 6330418K02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045821.1 | 3 | 604 | |||
| GenBank mRNA | AK005306.1 | - | 3 | 581 | ||
| AK031849.1 | - | 2 | 1134 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

6330418K02Rik| Gene Symbol: 6330418K02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045821.1 | 3 | 604 | |||
| GenBank mRNA | AK005306.1 | - | 3 | 581 | ||
| AK031849.1 | - | 2 | 1134 | |||