siRNA Details
| Gene Symbol: Arhgap27os3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045346.1 | 4 | 487 | |||
| GenBank mRNA | AK157183.1 | - | 4 | 489 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY752007.1 | - | 4 | 489 | Chr. 11 - 103344753 - 103348079 [+] on Build GRCm38 | |

Arhgap27os3
Genomic Map