siRNA Details
| Gene Symbol: Arhgap27os3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045346.1 | 4 | 1593 | |||
| GenBank mRNA | AK157183.1 | - | 4 | 1595 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arhgap27os3| Gene Symbol: Arhgap27os3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045346.1 | 4 | 1593 | |||
| GenBank mRNA | AK157183.1 | - | 4 | 1595 | ||