siRNA Details
| Gene Symbol: A630050E04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK041976.1 | - | 2 | 561 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A630050E04Rik| Gene Symbol: A630050E04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK041976.1 | - | 2 | 561 | ||