siRNA Details
Gene Symbol: A630050E04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK041976.1 | - | 1 | 392 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB638721.1 | - | 1 | 392 | Chr. 2 - 74100910 - 74102344 [-] on Build GRCm38 |
