siRNA Details
| Gene Symbol: 4930452G13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045060.1 | 2 | 283 | |||
| GenBank mRNA | AK015444.1 | - | 2 | 284 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714993.1 | - | 2 | 279 | Chr. 14 - 79702924 - 79712797 [+] on Build GRCm38 | |

4930452G13Rik
Genomic Map