siRNA Details
Gene Symbol: 4930452G13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045060.1 | 4 | 1030 | |||
GenBank mRNA | AK015444.1 | - | 4 | 1031 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930452G13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045060.1 | 4 | 1030 | |||
GenBank mRNA | AK015444.1 | - | 4 | 1031 |