siRNA Details
Gene Symbol: 4930452G13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045060.1 | 4 | 676 | |||
GenBank mRNA | AK015444.1 | - | 4 | 677 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714993.1 | - | 4 | 671 | Chr. 14 - 79702924 - 79712797 [+] on Build GRCm38 |
