siRNA Details
| Gene Symbol: 4930452G13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045060.1 | 4 | 745 | |||
| GenBank mRNA | AK015444.1 | - | 4 | 746 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930452G13Rik| Gene Symbol: 4930452G13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045060.1 | 4 | 745 | |||
| GenBank mRNA | AK015444.1 | - | 4 | 746 | ||