siRNA Details
| Gene Symbol: Gm21283 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_105058.1 | 1, 2 | 56 | |||
| GenBank mRNA | AK076897.1 | - | 1, 2 | 57 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm21283| Gene Symbol: Gm21283 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_105058.1 | 1, 2 | 56 | |||
| GenBank mRNA | AK076897.1 | - | 1, 2 | 57 | ||