siRNA Details
| Gene Symbol: Wdfy2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK158235.1 | - | 77 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY753019.1 | - | 1, 2 | 74 | Chr. 14 - 62837677 - 62838205 [-] on Build GRCm38 | |

Wdfy2
Genomic Map