siRNA Details
| Gene Symbol: Gm30025 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK147035.1 | - | 1 | 100 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY760904.1 | - | 1 | 45 | Chr. 12 - 80435262 - 80435931 [+] on Build GRCm38 | |

Gm30025
Genomic Map