siRNA Details
| Gene Symbol: Rassf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK142231.1 | - | 1 | 836 | ||
| BC004678.1 | - | 1 | 382 | |||
| BC025009.1 | - | 1 | 816 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rassf8| Gene Symbol: Rassf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK142231.1 | - | 1 | 836 | ||
| BC004678.1 | - | 1 | 382 | |||
| BC025009.1 | - | 1 | 816 | |||