siRNA Details
| Gene Symbol: 1700025K04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005576.1 | - | 1 | 18 | ||
| AK006351.1 | - | 44 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700025K04Rik| Gene Symbol: 1700025K04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005576.1 | - | 1 | 18 | ||
| AK006351.1 | - | 44 | ||||