siRNA Details
| Gene Symbol: 4933426I03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016932.1 | - | 1 | 228 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716612.1 | - | 1 | 228 | Chr. 10 - 82315649 - 82316559 [-] on Build GRCm38 | |

4933426I03Rik
Genomic Map