siRNA Details
| Gene Symbol: Spag17os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK035904.1 | - | 194 | |||
| AK047534.1 | - | 2 | 179 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spag17os| Gene Symbol: Spag17os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK035904.1 | - | 194 | |||
| AK047534.1 | - | 2 | 179 | |||