siRNA Details
| Gene Symbol: Dpp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK153591.1 | - | 2 | 2419 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dpp4| Gene Symbol: Dpp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK153591.1 | - | 2 | 2419 | ||