siRNA Details
Gene Symbol: Gm15239 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK080343.1 | - | 2 | 546 | ||
BB236368.2 | - | 2 | 569 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB749789.1 | - | 2 | 385 | Chr. X - 167361615 - 167364420 [+] on Build GRCm38 |
