siRNA Details
| Gene Symbol: Gm15239 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK080343.1 | - | 1 | 137 | ||
| BB236368.2 | - | 1 | 160 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB638905.1 | - | 1 | 137 | Chr. X - 167361455 - 167364416 [+] on Build GRCm38 | |

Gm15239
Genomic Map