siRNA Details
| Gene Symbol: Gm15239 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK080343.1 | - | 2 | 525 | ||
| BB236368.2 | - | 2 | 548 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm15239| Gene Symbol: Gm15239 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK080343.1 | - | 2 | 525 | ||
| BB236368.2 | - | 2 | 548 | |||