siRNA Details
| Gene Symbol: 9230112D13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020336.1 | - | 3 | 438 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY721146.1 | - | 3 | 436 | Chr. 14 - 34511822 - 34522800 [-] on Build GRCm38 | |

9230112D13Rik
Genomic Map