siRNA Details
| Gene Symbol: 4930571O06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110476.1 | 3 | 267 | |||
| GenBank mRNA | AK019805.1 | - | 3 | 268 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715894.1 | - | 3 | 268 | Chr. 8 - 113517282 - 113550810 [+] on Build GRCm38 | |

4930571O06Rik
Genomic Map