siRNA Details
| Gene Symbol: 2310034P14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK009626.1 | - | 1 | 123 | ||
| AV025514.1 | - | 1 | 137 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY709290.1 | - | 1 | 119 | Chr. 7 - 83884915 - 83885193 [+] on Build GRCm38 | |

2310034P14Rik
Genomic Map