siRNA Details
| Gene Symbol: 2310034O05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040679.1 | 3 | 492 | |||
| GenBank mRNA | AK009622.1 | - | 3 | 492 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2310034O05Rik| Gene Symbol: 2310034O05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040679.1 | 3 | 492 | |||
| GenBank mRNA | AK009622.1 | - | 3 | 492 | ||