siRNA Details
| Gene Symbol: Apbb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK009197.1 | - | 1 | 463 | ||
| BC022590.1 | - | 1 | 606 | |||
| BC038035.1 | - | 1 | 534 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Apbb2| Gene Symbol: Apbb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK009197.1 | - | 1 | 463 | ||
| BC022590.1 | - | 1 | 606 | |||
| BC038035.1 | - | 1 | 534 | |||