siRNA Details
| Gene Symbol: Fam179b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK132890.1 | - | 2 | 91 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CJ047139.1 | - | 2 | 91 | Chr. 12 - 64980630 - 64982936 [-] on Build GRCm38 | |

Fam179b
Genomic Map