siRNA Details
| Gene Symbol: Pggt1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK077685.1 | - | 1 | 624 | ||
| AK085164.1 | - | 1 | 1587 | |||
| AK131923.1 | - | 1 | 537 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pggt1b| Gene Symbol: Pggt1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK077685.1 | - | 1 | 624 | ||
| AK085164.1 | - | 1 | 1587 | |||
| AK131923.1 | - | 1 | 537 | |||