siRNA Details
| Gene Symbol: Dlx6os3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013501.1 | - | 1 | 219 | ||
| AK019329.1 | - | 1 | 219 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dlx6os3| Gene Symbol: Dlx6os3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013501.1 | - | 1 | 219 | ||
| AK019329.1 | - | 1 | 219 | |||