siRNA Details
| Gene Symbol: Gm12128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK037509.1 | - | 4 | 2871 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB154877.2 | - | 2 | 596 | Chr. 11 - 37671164 - 37673166 [+] on Build GRCm38 | |

Gm12128
Genomic Map