siRNA Details
| Gene Symbol: Gm12128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK037509.1 | - | 2 | 1467 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm12128| Gene Symbol: Gm12128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK037509.1 | - | 2 | 1467 | ||