siRNA Details
| Gene Symbol: Gm15446 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040366.1 | 1, 2 | 460 | |||
| GenBank mRNA | AK142681.1 | - | 1, 2 | 460 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY730939.1 | - | 1, 2 | 507 | Chr. 5 - 109933541 - 109940545 [+] on Build GRCm38 | |

Gm15446
Genomic Map