siRNA Details
| Gene Symbol: Gm15446 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040366.1 | 3 | 1331 | |||
| GenBank mRNA | AK142681.1 | - | 3 | 1331 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm15446| Gene Symbol: Gm15446 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040366.1 | 3 | 1331 | |||
| GenBank mRNA | AK142681.1 | - | 3 | 1331 | ||