siRNA Details
| Gene Symbol: Gm15446 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040366.1 | 3 | 599 | |||
| GenBank mRNA | AK142681.1 | - | 3 | 599 | ||
| Gene Symbol: Zfp932 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK009062.1 | - | 3 | 269 | ||
| AK032220.1 | - | 2 | 382 | |||
| BC012405.1 | - | 3 | 239 | |||

Gm15446
Genomic Map