siRNA Details
| Gene Symbol: Gm11587 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK135711.1 | - | 3 | 814 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB698797.1 | - | 2 | 194 | Chr. 1 - 61306644 - 61307746 [+] on Build GRCm38 | |

Gm11587
Genomic Map