siRNA Details
| Gene Symbol: 8430430B14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020236.1 | - | 2 | 244 | ||
| BY720584.1 | - | 2 | 241 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

8430430B14Rik| Gene Symbol: 8430430B14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020236.1 | - | 2 | 244 | ||
| BY720584.1 | - | 2 | 241 | |||