siRNA Details
| Gene Symbol: A330102I10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045073.1 | 2 | 190 | |||
| NR_045074.1 | 2 | 190 | ||||
| NR_045075.1 | 2 | 190 | ||||
| GenBank mRNA | AK020728.1 | - | 2 | 155 | ||
| BE655952.1 | - | 1 | 55 | |||
| BY284081.1 | - | 2 | 191 | |||
| CD769769.1 | - | 2 | 144 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY724424.1 | - | 2 | 154 | Chr. 13 - 29016291 - 29028689 [+] on Build GRCm38 | |

A330102I10Rik
Genomic Map