siRNA Details
| Gene Symbol: 1600025M17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038168.1 | 3 | 350 | |||
| GenBank mRNA | AA098421.1 | - | 3 | 349 | ||
| AK005539.1 | - | 3 | 305 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1600025M17Rik| Gene Symbol: 1600025M17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038168.1 | 3 | 350 | |||
| GenBank mRNA | AA098421.1 | - | 3 | 349 | ||
| AK005539.1 | - | 3 | 305 | |||