siRNA Details
| Gene Symbol: 3200001D21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045888.2 | 3 | 1471 | |||
| GenBank mRNA | AK014278.1 | - | 3 | 1472 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

3200001D21Rik| Gene Symbol: 3200001D21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045888.2 | 3 | 1471 | |||
| GenBank mRNA | AK014278.1 | - | 3 | 1472 | ||