siRNA Details
| Gene Symbol: Nhsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK041760.1 | - | 3, 4 | 393 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB637868.1 | - | 3, 4 | 390 | Chr. 10 - 18407640 - 18419525 [+] on Build GRCm38 | |

Nhsl1
Genomic Map